accessibility menu, dialog, popup

UserName

2H2H2HO2H2H2H2H2H

CAS RN 22739-76-0

IUPAC Name: (²H₇)propan-2-(²H)ol
Synonyms: (²H₇)propan-2-(²H)ol
Molecular Weight (g/mol): 68.15
Molecular Formula: C3H8O
InChi Key: KFZMGEQAYNKOFK-PIODKIDGSA-N
SMILES: [2H]OC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)