accessibility menu, dialog, popup

UserName

H2COOOOCH2

CAS RN 2274-11-5

IUPAC Name: 2-(prop-2-enoyloxy)ethyl prop-2-enoate
Synonyms: 2-(prop-2-enoyloxy)ethyl prop-2-enoate
Molecular Weight (g/mol): 170.16
Molecular Formula: C8H10O4
InChi Key: KUDUQBURMYMBIJ-UHFFFAOYSA-N
SMILES: C=CC(=O)OCCOC(=O)C=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)