accessibility menu, dialog, popup

UserName

CAS RN 2286-54-6

IUPAC Name: 3,3-diphenylpropanenitrile
Synonyms: 3,3-diphenylpropanenitrile
Molecular Weight (g/mol): 207.28
Molecular Formula: C15H13N
InChi Key: INERKLNEVAZSCI-UHFFFAOYSA-N
SMILES: N#CCC(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results