CAS RN 22918-01-0
IUPAC Name:
2-bromo-4-chloropyridine
Synonyms:
2-bromo-4-chloropyridine
Molecular Weight (g/mol):
192.44
Molecular Formula:
C5H3BrClN
InChi Key:
SURKZMFXICWLHU-UHFFFAOYSA-N
SMILES:
ClC1=CC=NC(Br)=C1
1
–
1
of
1
results
1
2-Bromo-4-chloropyridine, 97%
CAS: 22918-01-0 Molecular Formula: C5H3BrClN Molecular Weight (g/mol): 192.44 MDL Number: MFCD03840757 InChI Key: SURKZMFXICWLHU-UHFFFAOYSA-N Synonym: 2-bromo-4-chloro-pyridine,pyridine, 2-bromo-4-chloro,pubchem13533,acmc-1cled,2-brom-4-chlorpyridin;,2-bromo-4 chloropyridine,2-bromo-4-chloro pyridine,2-bromo-4-chloropyridine;,2-bromo-4-pyridyl chloride,2-bromo-4-pyridyl chloride; PubChem CID: 2762836 IUPAC Name: 2-bromo-4-chloropyridine SMILES: ClC1=CC=NC(Br)=C1