accessibility menu, dialog, popup

UserName

BrO

CAS RN 23145-07-5

IUPAC Name: 5-bromo-1-benzofuran
Synonyms: 5-bromo-1-benzofuran
Molecular Weight (g/mol): 197.03
Molecular Formula: C8H5BrO
InChi Key: AYOVPQORFBWFNO-UHFFFAOYSA-N
SMILES: BrC1=CC=C2OC=CC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)