accessibility menu, dialog, popup

UserName

CAS RN 232941-00-3

IUPAC Name: 3-(1,3-benzothiazol-2-yl)-4-methylthiophen-2-amine
Synonyms: 3-(1,3-benzothiazol-2-yl)-4-methylthiophen-2-amine
Molecular Weight (g/mol): 246.35
Molecular Formula: C12H10N2S2
InChi Key: YXXFAQKMMOVVIA-UHFFFAOYSA-N
SMILES: CC1=CSC(N)=C1C1=NC2=CC=CC=C2S1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results