accessibility menu, dialog, popup

UserName

ClNSN

CAS RN 23576-81-0

IUPAC Name: 6-chloroimidazo[2,1-b][1,3]thiazole
Synonyms: 6-chloroimidazo[2,1-b][1,3]thiazole
Molecular Weight (g/mol): 158.6
Molecular Formula: C5H3ClN2S
InChi Key: WOZMDYAJHVHPMD-UHFFFAOYSA-N
SMILES: ClC1=CN2C=CSC2=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results