accessibility menu, dialog, popup

UserName

CH3CH3OCH3CH3OCu

CAS RN 23578-23-6

IUPAC Name: copper(2+) bis(propan-2-olate)
Synonyms: copper(2+) bis(propan-2-olate)
Molecular Weight (g/mol): 181.72
Molecular Formula: C6H14CuO2
InChi Key: VNGORJHUDAPOQZ-UHFFFAOYSA-N
SMILES: [Cu++].CC(C)[O-].CC(C)[O-]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results