accessibility menu, dialog, popup

UserName

NH2N

CAS RN 2359-60-6

IUPAC Name: 4-(piperidin-1-yl)aniline
Synonyms: 4-(piperidin-1-yl)aniline
Molecular Weight (g/mol): 176.26
Molecular Formula: C11H16N2
InChi Key: TVOSOIXYPHKEAR-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)N1CCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results