accessibility menu, dialog, popup

UserName

NNH2

CAS RN 23687-27-6

IUPAC Name: isoquinolin-8-amine
Synonyms: isoquinolin-8-amine
Molecular Weight (g/mol): 144.18
Molecular Formula: C9H8N2
InChi Key: GUSYANXQYUJOBH-UHFFFAOYSA-N
SMILES: NC1=C2C=NC=CC2=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results