CAS RN 2370-61-8
CAS RN 2370-61-8
IUPAC Name:
2-amino-3-(2-hydroxyphenyl)propanoic acid
Synonyms:
2-tyrosine o-tyrosine o-tyrosine, DL- DL-o-tyrosine DL- O-tyrosine ortho-tyrosine
Molecular Weight (g/mol):
181.19
Molecular Formula:
C9H11NO3
InChi Key:
WRFPVMFCRNYQNR-UHFFFAOYNA-N
SMILES:
NC(CC1=CC=CC=C1O)C(O)=O
1
–
1
of
1
results
1
DL-o-Tyrosine, 98%, Thermo Scientific™
CAS: 2370-61-8 Molecular Formula: C9H11NO3 Molecular Weight (g/mol): 181.19 MDL Number: MFCD00021725 InChI Key: WRFPVMFCRNYQNR-UHFFFAOYNA-N Synonym: dl-o-tyrosine,2-amino-3-2-hydroxyphenyl propanoic acid,o-tyrosine,2-hydroxyphenylalanine,2-hydroxy-dl-phenylalanine,ortho-tyrosine,2-tyrosine,o-tyrosine, dl,dl-3-o-hydroxyphenyl alanine,2-amino-3-2-hydroxyphenyl propionic acid PubChem CID: 91482 IUPAC Name: 2-amino-3-(2-hydroxyphenyl)propanoic acid SMILES: NC(CC1=CC=CC=C1O)C(O)=O