accessibility menu, dialog, popup

UserName

CAS RN 2386-56-3

IUPAC Name: potassium methanesulfonate
Synonyms: potassium methanesulfonate
Molecular Weight (g/mol): 134.19
Molecular Formula: CH3KO3S
InChi Key: XWIJIXWOZCRYEL-UHFFFAOYSA-M
SMILES: [K+].CS([O-])(=O)=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results