CAS RN 23877-12-5
IUPAC Name:
tert-butyl 2-bromo-2-methylpropanoate
Synonyms:
tert-butyl 2-bromo-2-methylpropanoate
Molecular Weight (g/mol):
223.11
Molecular Formula:
C8H15BrO2
InChi Key:
IGVNJALYNQVQIT-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)C(C)(C)Br
1
–
4
of
4
results
1
tert-Butyl 2-bromoisobutyrate, 97%
CAS: 23877-12-5 Molecular Formula: C8H15BrO2 Molecular Weight (g/mol): 223.11 MDL Number: MFCD00051908 InChI Key: IGVNJALYNQVQIT-UHFFFAOYSA-N PubChem CID: 90290 IUPAC Name: tert-butyl 2-bromo-2-methylpropanoate SMILES: CC(C)(C)OC(=O)C(C)(C)Br
2
tert-Butyl 2-bromo-2-methylpropanoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
tert-Butyl 2-bromo-2-methylpropanoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
tert-Butyl 2-bromo-2-methylpropanoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.