accessibility menu, dialog, popup

UserName

CAS RN 23911-26-4

OHONNOOONOOO

CAS RN 23911-26-4

IUPAC Name: 2-{bis[2-(2,6-dioxomorpholin-4-yl)ethyl]amino}acetic acid
Synonyms: {bis[2-(2,6-dioxomorpholin-4-yl)ethyl]amino}acetic acid
Molecular Weight (g/mol): 357.32
Molecular Formula: C14H19N3O8
InChi Key: RAZLJUXJEOEYAM-UHFFFAOYSA-N
SMILES: OC(=O)CN(CCN1CC(=O)OC(=O)C1)CCN1CC(=O)OC(=O)C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results