CAS RN 2396-61-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3,3'-Oxydipropanol, TRC
CAS: 2396-61-4 Molecular Formula: C6H14O3 Molecular Weight (g/mol): 134.17 Synonym: 3,3'-Oxydi-1-propanol,3,3'-Oxybis[1-propanol],3,3'-Dihydroxydipropyl Ether,4-Oxa-1,7-heptanediol,Bis(3-hydroxypropyl) Ether,Di-n-propylene Glycol,NSC 362072 IUPAC Name: 3-(3-hydroxypropoxy)propan-1-ol SMILES: OCCCOCCCO