CAS RN 2396-78-3
CAS RN 2396-78-3
IUPAC Name:
methyl (3E)-hex-3-enoate
Synonyms:
methyl (3E)-hex-3-enoate
Molecular Weight (g/mol):
128.17
Molecular Formula:
C7H12O2
InChi Key:
XEAIHUDTEINXFG-SNAWJCMRSA-N
SMILES:
CC\C=C\CC(=O)OC
1
–
1
of
1
results
1
Methyl 3-hexenoate, 95+%, predominantly trans, Thermo Scientific™
CAS: 2396-78-3 Molecular Formula: C7H12O2 Molecular Weight (g/mol): 128.17 MDL Number: MFCD00672771 InChI Key: XEAIHUDTEINXFG-SNAWJCMRSA-N Synonym: 3-hexenoic acid, methyl ester,methyl 3-hexenoate,methyl trans-3-hexenoate,methyl e-hex-3-enoate,honeyflor,3-hexenoic acid, methyl ester, e,unii-7ip0t834pb,methyl hex-3-enoate,methyl e-3-hexenoate,methyl trans 3 hexenoate PubChem CID: 5362782 IUPAC Name: methyl (E)-hex-3-enoate SMILES: CCC=CCC(=O)OC