CAS RN 24003-58-5
CAS RN 24003-58-5
IUPAC Name:
{[(2,6-dimethylphenyl)carbamoyl]methyl}triethylazanium bromide
Synonyms:
{[(2,6-dimethylphenyl)carbamoyl]methyl}triethylazanium bromide
Molecular Weight (g/mol):
343.31
Molecular Formula:
C16H27BrN2O
InChi Key:
DLHMKHREUTXMCH-UHFFFAOYSA-N
SMILES:
[Br-].CC[N+](CC)(CC)CC(=O)NC1=C(C)C=CC=C1C