accessibility menu, dialog, popup

UserName

CAS RN 2404-55-9

IUPAC Name: trimethyl-λ⁵-phosphanethione
Synonyms: trimethyl-λ⁵-phosphanethione
Molecular Weight (g/mol): 108.14
Molecular Formula: C3H9PS
InChi Key: KTFAZNVGJUIWJM-UHFFFAOYSA-N
SMILES: CP(C)(C)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results