accessibility menu, dialog, popup

UserName

CAS RN 2412-80-8

H3COOCH3CH3

CAS RN 2412-80-8

IUPAC Name: methyl 4-methylpentanoate
Synonyms: methyl methylvalerate
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: KBCOVKHULBZKNY-UHFFFAOYSA-N
SMILES: COC(=O)CCC(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results