CAS RN 24234-92-2
CAS RN 24234-92-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-[4-Chloro-2-(2-chlorobenzoyl)phenyl]-2-[(2-hydroxyethyl)amino]acetamide, TRC
CAS: 24234-92-2 Molecular Formula: C17H16Cl2N2O3 Molecular Weight (g/mol): 367.23 Synonym: 4'-Chloro-2'-(o-chlorobenzoyl)-2-[(2-hydroxyethyl)amino]acetamide,N-[4-Chloro-2-(2-chlorobenzoyl)phenyl]-2-[(2-hydroxyethyl)amino]acetamide IUPAC Name: N-[4-chloro-2-(2-chlorobenzoyl)phenyl]-2-(2-hydroxyethylamino)acetamide SMILES: OCCNCC(=O)Nc1ccc(Cl)cc1C(=O)c2ccccc2Cl