accessibility menu, dialog, popup

UserName

CAS RN 243641-37-4

H3CNOOCH3H3CCH3N

CAS RN 243641-37-4

IUPAC Name: tert-butyl 7-methyl-1,2,3,4-tetrahydro-1,8-naphthyridine-1-carboxylate
Synonyms: tert-butyl 7-methyl-3,4-dihydro-2H-1,8-naphthyridine-1-carboxylate
Molecular Weight (g/mol): 248.33
Molecular Formula: C14H20N2O2
InChi Key: CHWMFCCGEWQABK-UHFFFAOYSA-N
SMILES: CC1=CC=C2CCCN(C(=O)OC(C)(C)C)C2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)