CAS RN 243669-48-9
IUPAC Name:
ethyl 5'-amino-[2,3'-bithiophene]-4'-carboxylate
Synonyms:
ethyl 5'-amino-[2,3'-bithiophene]-4'-carboxylate
Molecular Weight (g/mol):
253.33
Molecular Formula:
C11H11NO2S2
InChi Key:
AKFDBWIUWIWHRK-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=C(N)SC=C1C1=CC=CS1
1
–
1
of
1
results
1
Ethyl 2-amino-4-(2-thienyl)thiophene-3-carboxylate, 97%, Thermo Scientific™
CAS: 243669-48-9 Molecular Formula: C11H11NO2S2 Molecular Weight (g/mol): 253.33 MDL Number: MFCD00115120 InChI Key: AKFDBWIUWIWHRK-UHFFFAOYSA-N Synonym: ethyl 2-amino-4-2-thienyl thiophene-3-carboxylate,ethyl 5'-amino-2,3'-bithiophene-4'-carboxylate,ethyl 2-amino-4-thiophen-2-yl thiophene-3-carboxylate,5'-amino-2,3' bithiophenyl-4'-carboxylic acid ethyl ester,ethyl 5'-amino 2,3'-bithiophene-4'-carboxylate,ethyl 2-azanyl-4-thiophen-2-yl-thiophene-3-carboxylate,2,3'-bithiophene-4'-carboxylicacid, 5'-amino-, ethyl ester,2-amino-4-thiophen-2-yl-3-thiophenecarboxylic acid ethyl ester,maybridge1_003774 PubChem CID: 718227 SMILES: CCOC(=O)C1=C(N)SC=C1C1=CC=CS1