CAS RN 24412-91-7
CAS RN 24412-91-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Glycidyl Caprylate, TRC
CAS: 24412-91-7 Molecular Formula: C11H20O3 Molecular Weight (g/mol): 200.27 Synonym: Octanoic Acid 2,3-Epoxypropyl Ester,Octanoic Acid Oxiranylmethyl Ester,2,3-Epoxy-1-propanol Octanoate,2,3-Epoxypropyl Octanoate IUPAC Name: oxiran-2-ylmethyl octanoate SMILES: CCCCCCCC(=O)OCC1CO1