accessibility menu, dialog, popup

UserName

CAS RN 24422-15-9

SClClOClCl

CAS RN 24422-15-9

IUPAC Name: trichlorothiophene-2-carbonyl chloride
Synonyms: trichlorothiophene-2-carbonyl chloride
Molecular Weight (g/mol): 249.91
Molecular Formula: C5Cl4OS
InChi Key: ASESVIHJSOBINF-UHFFFAOYSA-N
SMILES: ClC(=O)C1=C(Cl)C(Cl)=C(Cl)S1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)