accessibility menu, dialog, popup

UserName

H3COOCH3H3CCH3Br

CAS RN 24457-21-4

IUPAC Name: tert-butyl 2-bromobutanoate
Synonyms: tert-butyl 2-bromobutanoate
Molecular Weight (g/mol): 223.11
Molecular Formula: C8H15BrO2
InChi Key: RIUJWUWLGXBICR-UHFFFAOYNA-N
SMILES: CCC(Br)C(=O)OC(C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results