CAS RN 25470-96-6
CAS RN 25470-96-6
IUPAC Name:
2,4-dioxopentan-3-ide methanidylidyneoxidanium triphenylphosphane rhodium
Synonyms:
carbon monoxide triphenylphosphine acetylacetonate rhodium
Molecular Weight (g/mol):
492.32
Molecular Formula:
C24H22O3PRh
InChi Key:
ZETXURAVPSJSPU-UHFFFAOYSA-N
SMILES:
[Rh].[C-]#[O+].CC(=O)[CH-]C(C)=O.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Carbonyl-2,4-pentanedionato(triphenylphosphine)rhodium(I), Rh 20%
CAS: 25470-96-6 Molecular Formula: C24H22O3PRh Molecular Weight (g/mol): 492.32 MDL Number: MFCD00064611 InChI Key: UGUXXRZSXZLSCY-MECAPONASA-M IUPAC Name: λ¹-rhodium(1+) methanidylidyneoxidanium triphenylphosphane (2Z)-4-oxopent-2-en-2-olate SMILES: [Rh+].[C-]#[O+].C\C([O-])=C\C(C)=O.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1