accessibility menu, dialog, popup

UserName

H3CNNOCH3

CAS RN 25680-58-4

IUPAC Name: 2-ethyl-3-methoxypyrazine
Synonyms: 2-ethyl-3-methoxypyrazine
Molecular Weight (g/mol): 138.17
Molecular Formula: C7H10N2O
InChi Key: DPCILIMHENXHQX-UHFFFAOYSA-N
SMILES: CCC1=NC=CN=C1OC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results