accessibility menu, dialog, popup

UserName

CAS RN 25842-32-4

NNH2

CAS RN 25842-32-4

IUPAC Name: 2-(4-benzylpiperidin-1-yl)ethan-1-amine
Synonyms: 2-(4-benzylpiperidin-1-yl)ethanamine
Molecular Weight (g/mol): 218.34
Molecular Formula: C14H22N2
InChi Key: PCNDXYHWLSHXMV-UHFFFAOYSA-N
SMILES: NCCN1CCC(CC2=CC=CC=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results