accessibility menu, dialog, popup

UserName

ONOO

CAS RN 25891-31-0

IUPAC Name: N,N-diformylformamide
Synonyms: N,N-diformylformamide
Molecular Weight (g/mol): 101.06
Molecular Formula: C3H3NO3
InChi Key: PFBAGGWJGZAGCG-UHFFFAOYSA-N
SMILES: O=CN(C=O)C=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results