CAS RN 2592-05-4
IUPAC Name:
N-{[(3Z)-3H-indol-3-ylidene]methyl}hydroxylamine
Synonyms:
N-[(3Z)-indol-3-ylidenemethyl]hydroxylamine
Molecular Weight (g/mol):
160.18
Molecular Formula:
C9H8N2O
InChi Key:
HFNHXOHLZXORGW-VOTSOKGWSA-N
SMILES:
ON\C=C1/C=NC2=CC=CC=C12
1
–
1
of
1
results
1
Indole-3-carboxaldoxime, 97%, Thermo Scientific™
CAS: 2592-05-4 MDL Number: MFCD00022737