accessibility menu, dialog, popup

UserName

H3COPONOOOCH3

CAS RN 2609-49-6

IUPAC Name: diethyl [(4-nitrophenyl)methyl]phosphonate
Synonyms: diethyl (4-nitrophenyl)methylphosphonate
Molecular Weight (g/mol): 273.23
Molecular Formula: C11H16NO5P
InChi Key: FORMFFDDQMCTCT-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC1=CC=C(C=C1)[N+]([O-])=O)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results