accessibility menu, dialog, popup

UserName

WOOOOOH3CCH3H3CCH3CH3

CAS RN 26143-11-3

IUPAC Name: pentaethanolate tungsten
Synonyms: pentakis(ethoxide) tungsten
Molecular Weight (g/mol): 409.15
Molecular Formula: C10H25O5W
InChi Key: RBZKAYBXYBXUCL-UHFFFAOYSA-N
SMILES: [W].CC[O-].CC[O-].CC[O-].CC[O-].CC[O-]
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results