accessibility menu, dialog, popup

UserName

ClClNHONH2(E/Z)

CAS RN 261761-55-1

IUPAC Name: 2,3-dichloro-N'-hydroxybenzene-1-carboximidamide
Synonyms: 2,3-dichloro-N'-hydroxybenzenecarboximidamide
Molecular Weight (g/mol): 205.04
Molecular Formula: C7H6Cl2N2O
InChi Key: OPOPCNCEDLBYPV-UHFFFAOYSA-N
SMILES: NC(=NO)C1=C(Cl)C(Cl)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results