accessibility menu, dialog, popup

UserName

CAS RN 26218-80-4

CH3OOOCH3N

CAS RN 26218-80-4

IUPAC Name: methyl 6-methoxypyridine-3-carboxylate
Synonyms: methyl 6-methoxypyridine-3-carboxylate
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: OVLDWZNVBDRZNN-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(OC)N=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results