accessibility menu, dialog, popup

UserName

OI

CAS RN 26272-85-5

IUPAC Name: 3-iodooxetane
Synonyms: 3-iodooxetane
Molecular Weight (g/mol): 183.98
Molecular Formula: C3H5IO
InChi Key: KBEIFKMKVCDETC-UHFFFAOYSA-N
SMILES: IC1COC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results