accessibility menu, dialog, popup

UserName

CAS RN 26351-19-9

NHOONHOHNOH2O

CAS RN 26351-19-9

IUPAC Name: 5-(hydroxyimino)-1,3-diazinane-2,4,6-trione hydrate
Synonyms: 5-(hydroxyimino)-1,3-diazinane-2,4,6-trione hydrate
Molecular Weight (g/mol): 175.1
Molecular Formula: C4H5N3O5
InChi Key: YHAIHNZKUCGXRI-UHFFFAOYSA-N
SMILES: O.ON=C1C(=O)NC(=O)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results