accessibility menu, dialog, popup

UserName

OH2NCl

CAS RN 26378-53-0

IUPAC Name: 2-(2-chlorophenoxy)ethan-1-amine
Synonyms: 2-(2-chlorophenoxy)ethanamine
Molecular Weight (g/mol): 171.62
Molecular Formula: C8H10ClNO
InChi Key: NAPNYPMMDVRKGM-UHFFFAOYSA-N
SMILES: NCCOC1=CC=CC=C1Cl
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)