accessibility menu, dialog, popup

UserName

OI

CAS RN 264617-03-0

IUPAC Name: 7-iodo-2,3-dihydro-1-benzofuran
Synonyms: 7-iodo-2,3-dihydro-1-benzofuran
Molecular Weight (g/mol): 246.05
Molecular Formula: C8H7IO
InChi Key: IASDAJCCAFGOCE-UHFFFAOYSA-N
SMILES: IC1=CC=CC2=C1OCC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results