accessibility menu, dialog, popup

UserName

CAS RN 26510-95-2

H3COOOBr

CAS RN 26510-95-2

IUPAC Name: ethyl 3-(4-bromophenyl)-3-oxopropanoate
Synonyms: ethyl 3-(4-bromophenyl)-3-oxopropanoate
Molecular Weight (g/mol): 271.11
Molecular Formula: C11H11BrO3
InChi Key: PBDYXCKRDRCJDC-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)C1=CC=C(Br)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results