accessibility menu, dialog, popup

UserName

CH3OOONH

CAS RN 26518-71-8

IUPAC Name: 6-acetyl-3,4-dihydro-2H-1,4-benzoxazin-3-one
Synonyms: 6-acetyl-2,4-dihydro-1,4-benzoxazin-3-one
Molecular Weight (g/mol): 191.19
Molecular Formula: C10H9NO3
InChi Key: BKJFWHFUERNXLJ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C2OCC(=O)NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results