accessibility menu, dialog, popup

UserName

OSOOOKn

CAS RN 26837-42-3

IUPAC Name: potassium ethenyl sulfate
Synonyms: potassium ethenyl sulfate
Molecular Weight (g/mol): 162.2
Molecular Formula: C2H3KO4S
InChi Key: WZAPMUSQALINQD-UHFFFAOYSA-M
SMILES: [K+].[O-]S(=O)(=O)OC=C
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)