accessibility menu, dialog, popup

UserName

CAS RN 2689-68-1

CH3OOSO

CAS RN 2689-68-1

IUPAC Name: methyl 4-oxothiolane-3-carboxylate
Synonyms: methyl 4-oxothiolane-3-carboxylate
Molecular Weight (g/mol): 160.19
Molecular Formula: C6H8O3S
InChi Key: LEAKUJFYXNILRB-UHFFFAOYNA-N
SMILES: COC(=O)C1CSCC1=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results