accessibility menu, dialog, popup

UserName

ON

CAS RN 271-95-4

IUPAC Name: 1,2-benzoxazole
Synonyms: benzisoxazole indoxazene benz(d)isoxazole 4,5-benzisoxazole
Molecular Weight (g/mol): 119.12
Molecular Formula: C7H5NO
InChi Key: KTZQTRPPVKQPFO-UHFFFAOYSA-N
SMILES: O1N=CC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)