accessibility menu, dialog, popup

UserName

NHOSOOHOMeMen

CAS RN 27119-07-9

IUPAC Name: 2-methyl-2-(prop-2-enamido)propane-1-sulfonic acid
Synonyms: polyacrylamidomethylpropane sulfonic acid
Molecular Weight (g/mol): 207.24
Molecular Formula: C7H13NO4S
InChi Key: XHZPRMZZQOIPDS-UHFFFAOYSA-N
SMILES: CC(C)(CS(O)(=O)=O)NC(=O)C=C
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)