accessibility menu, dialog, popup

UserName

ON

CAS RN 272-62-8

IUPAC Name: furo[3,2-b]pyridine
Synonyms: furo[3,2-b]pyridine
Molecular Weight (g/mol): 119.12
Molecular Formula: C7H5NO
InChi Key: YRTCKZIKGWZNCU-UHFFFAOYSA-N
SMILES: O1C=CC2=NC=CC=C12
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results