accessibility menu, dialog, popup

UserName

CAS RN 274-56-6

NN

CAS RN 274-56-6

IUPAC Name: pyrazolo[1,5-a]pyridine
Synonyms: pyrazolo[1,5-a]pyridine
Molecular Weight (g/mol): 118.14
Molecular Formula: C7H6N2
InChi Key: DVUBDHRTVYLIPA-UHFFFAOYSA-N
SMILES: C1=NN2C=CC=CC2=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results