CAS RN 274257-34-0
IUPAC Name:
potassium trifluoro(2-methylphenyl)boranuide
Synonyms:
potassium trifluoro(2-methylphenyl)boranuide
Molecular Weight (g/mol):
198.04
Molecular Formula:
C7H7BF3K
InChi Key:
ROMUTKWSUQKVLS-UHFFFAOYSA-N
SMILES:
[K+].CC1=CC=CC=C1[B-](F)(F)F
1
–
1
of
1
results
1
Potassium 2-methylphenyltrifluoroborate, 96%, Thermo Scientific™
CAS: 274257-34-0 Molecular Formula: C7H7BF3K Molecular Weight (g/mol): 198.04 MDL Number: MFCD03790006 InChI Key: ROMUTKWSUQKVLS-UHFFFAOYSA-N Synonym: potassium trifluoro o-tolyl borate,potassium 2-methylphenyl trifluoroborate,potassium o-tolyltrifluoroborate,potassium trifluoro 2-methylphenyl boranuide,potassium 2-methylphenyltrifluoroborate,pubchem11432,potassiumo-tolyltrifluoroborate,trifluoro 2-methylphenyl potassioboron v,potassium trifluoro-2-methylphenyl boranuide,potassium trifluoro 2-methylphenyl borate 1- PubChem CID: 23674703 IUPAC Name: potassium;trifluoro-(2-methylphenyl)boranuide SMILES: [K+].CC1=CC=CC=C1[B-](F)(F)F