CAS RN 2742962-60-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R)-N-Benzyl-3-methoxy-2-(N-methylacetamido)propanamide, TRC
CAS: 2742962-60-1 Molecular Formula: C14H20N2O3 Molecular Weight (g/mol): 264.32 Synonym: (R)-N-Benzyl-2-[N-acetyl N-Methyl-amino]-3-methoxypropionamide; IUPAC Name: (2R)-2-[acetyl(methyl)amino]-N-benzyl-3-methoxypropanamide SMILES: O=C([C@H](N(C(C)=O)C)COC)NCC1=CC=CC=C1