CAS RN 27460-02-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dodecyl diphenyl phosphate, TRC
CAS: 27460-02-2 Molecular Formula: C24 H35 O4 P Molecular Weight (g/mol): 418.506 Synonym: Diphenyl lauryl phosphate,Dodecyl diphenyl phosphate IUPAC Name: dodecyl diphenyl phosphate SMILES: CCCCCCCCCCCCOP(=O)(Oc1ccccc1)Oc2ccccc2