accessibility menu, dialog, popup

UserName

HNHO

CAS RN 27640-31-9

IUPAC Name: (2,3-dihydro-1H-indol-2-yl)methanol
Synonyms: 2,3-dihydro-1H-indol-2-ylmethanol
Molecular Weight (g/mol): 149.19
Molecular Formula: C9H11NO
InChi Key: GRPOFAKYHPAXNP-UHFFFAOYNA-N
SMILES: OCC1CC2=CC=CC=C2N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results